missing translation for 'onlineSavingsMsg'
Learn More

Daunomycin hydrochloride, MP Biomedicals™

Product Code. 11432172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432172 1 mg 1mg
11442172 5 mg 5mg
11462172 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432172 Supplier MP Biomedicals Supplier No. 0215077701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Photostability, Antileukemic, and drug metabolism studies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23541-50-6
Synonym Daunorubicin hydrochloride, Leukaemomycin C

Specifications

Type Daunomycin Hydrochloride
Physical Form Solid
Quantity 1 mg
Molecular Formula C27H30ClNO10
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Molecular Weight (g/mol) 563.984
PubChem CID 62770
Formula Weight 563.99
Chemical Name or Material Daunomycin hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.