accessibility menu, dialog, popup

UserName

ClH3COOOH(S)OH(S)O(R)(S)NH2(S)OH(S)CH3OOHOCH3OH

CAS RN 23541-50-6

IUPAC Name: hydrogen (8S,10S)-8-acetyl-10-{[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy}-6,8,11-trihydroxy-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
Synonyms: hydrogen daunorubicin chloride
Molecular Weight (g/mol): 563.98
Molecular Formula: C27H30ClNO10
InChi Key: GUGHGUXZJWAIAS-QQYBVWGSSA-N
SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3O[C@H]3C[C@H](N)[C@H](O)[C@H](C)O3)C(C)=O)C(O)=C1C2=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (4)
Type 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)