missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dapagliflozin

Product Code. 16564274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16564274 5 g 5g
16554274 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16564274 Supplier Thermo Scientific Chemicals Supplier No. 464620050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 461432-26-8 | C21H25ClO6 | 408.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461432-26-8
Molecular Formula C21H25ClO6
Molecular Weight (g/mol) 408.88
InChI Key JVHXJTBJCFBINQ-UHFFFAOYNA-N
IUPAC Name 2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1

Specifications

Melting Point 65.0°C to 70.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol
Formula Weight 408.88
Chemical Name or Material Dapagliflozin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.