accessibility menu, dialog, popup

UserName

CAS RN 461432-26-8

CH3OOOHOHOHOHCl

CAS RN 461432-26-8

IUPAC Name: 2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: dapagliflozin
Molecular Weight (g/mol): 408.88
Molecular Formula: C21H25ClO6
InChi Key: JVHXJTBJCFBINQ-UHFFFAOYNA-N
SMILES: CCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results