Learn More
Dantrolene sodium hemiheptahydrate, Thermo Scientific™
Product Code. 17740679 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17740679 5 g 5g
17750679 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17740679 Supplier Thermo Scientific Acros Supplier No. 469940050
Request Bulk or Custom Format
Specifications
Chemical Name or Material Dantrolene sodium hemiheptahydrate
CAS 24868-20-0
Synonym 1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt
Quantity 5 g
Solubility slightly soluble aq. soln.
Melting Point 225°C to 226°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C14 H9 N4 Na O5 · 3.5 H2O
Physical Form Crystalline Powder
Color Yellow to Orange to Brown
Purity ≥97.5%