missing translation for 'onlineSavingsMsg'
Learn More

D-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. p-7040761
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437108 1 g 1g
15447108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437108 Supplier Thermo Scientific Alfa Aesar Supplier No. H60622.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.

Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14907-27-8
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00038992
InChI Key XNFNGGQRDXFYMM-UHFFFAOYNA-N
Synonym d-tryptophan methyl ester hydrochloride, h-d-trp-ome.hcl, r-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl 2r-2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl d-tryptophanate hydrochloride, d-tryptophan methyl ester hcl, h-d-trp-ome hcl, dtrp-ome.hcl, pubchem10879, ksc174m8r
PubChem CID 11448200
IUPAC Name methyl (2R)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 213°C to 216°C
Solubility Information Soluble in water (150mg/ml),dimethyl sulfoxide and methanol (50mg/ml).
Quantity 1 g
Formula Weight 254.71
Optical Rotation −18° (c=5 in Methanol)
Percent Purity 98%
Chemical Name or Material D-Tryptophan methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.