accessibility menu, dialog, popup

UserName

CAS RN 14907-27-8

NH(R)ONH2OCH3HCl

CAS RN 14907-27-8

IUPAC Name: methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Synonyms: methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Molecular Weight (g/mol): 254.71
Molecular Formula: C12H15ClN2O2
InChi Key: XNFNGGQRDXFYMM-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)