Learn More
D-Threose (~0.4 M aqueous solution), TRC
Product Code. 30453430
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453430 1 g 1g
30451328 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30453430 Supplier Toronto Research Chemicals Supplier No. TRCT4055101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-43-2
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.11
Synonym (2S,3R)-2,3,4-Trihydroxy-butanal, [S-(R*,S*)]-2,3,4,-Trihydroxy-butanal, D-(-)-Threose
IUPAC Name (2S,3R)-2,3,4-trihydroxybutanal
SMILES OC[C@@H](O)[C@H](O)C=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C4H8O4/c5-1-3(7)4(8)2-6/h1,3-4,6-8H,2H2/t3-,4-/m1/s1
Formula Weight 120.0423
Chemical Name or Material D-Threose