accessibility menu, dialog, popup

UserName

OH(R)OHO(S)HO

CAS RN 95-43-2

IUPAC Name: (3S,4R)-oxolane-2,3,4-triol
Synonyms: D-threo-tetrose
Molecular Weight (g/mol): 120.1
Molecular Formula: C4H8O4
InChi Key: FMAORJIQYMIRHF-BCDHYOAOSA-N
SMILES: O[C@@H]1COC(O)[C@H]1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results