missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Melibiose has been used in a study to improve the preparation of benzoylated ethyl 1-thioglycosides.
- It has also been used in a study to investigate the effect of water plasticization on the molecular mobility and crystallization tendency of amorphous disaccharides.
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Optical Rotation | +135 ± 10° (c=4, water) |
| Formula Weight | 360.3 |
| Percent Purity | 99.9% |
| Physical Form | Powder |
| Chemical Name or Material | α-D-(+)-Melibiose monohydrate |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?