accessibility menu, dialog, popup

UserName

HOOOOOHOHOHHOOHHOHO

CAS RN 585-99-9

IUPAC Name: 6-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Synonyms: polydextrose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: DLRVVLDZNNYCBX-UHFFFAOYNA-N
SMILES: OCC1OC(OCC2OC(O)C(O)C(O)C2O)C(O)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)