missing translation for 'onlineSavingsMsg'
Learn More

D-Mannosamine Hydrochloride, TRC

Product Code. 30012432
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012432 1 g 1g
30010652 100 mg 100mg
30004998 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012432 Supplier Toronto Research Chemicals Supplier No. TRCM1670001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5505-63-5
Molecular Formula C6 H13 N O5 . Cl H
Molecular Weight (g/mol) 215.63
IUPAC Name (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride
SMILES Cl.N[C@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4-,5-,6-;/m1./s1
Formula Weight 215.0561
Chemical Name or Material D-Mannosamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.