CAS RN 5505-63-5
CAS RN 5505-63-5
IUPAC Name:
hydrogen (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanoic acid chloride
Synonyms:
hydrogen D-mannosaminic acid chloride
Molecular Weight (g/mol):
231.63
Molecular Formula:
C6H14ClNO6
InChi Key:
JMVMHPCPPSZGNY-BZWNWTOPSA-N
SMILES:
[H+].[Cl-].N[C@@H]([C@@H](O)[C@H](O)[C@H](O)CO)C(O)=O
1
–
3
of
3
results
1
Thermo Scientific Chemicals D-Mannosamine hydrochloride, 98%
CAS: 5505-63-5 Molecular Formula: C6H14ClNO5 Molecular Weight (g/mol): 215.63 MDL Number: MFCD00064557 InChI Key: CBOJBBMQJBVCMW-DEZHIRTDSA-N Synonym: 2-amino-2-deoxy-d-mannosehydrochloride PubChem CID: 133109008 IUPAC Name: (2R,3S,4R,5S)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride SMILES: C(C(C(C(C(C=O)N)O)O)O)O.Cl
3
D-Mannosamine Hydrochloride, TRC
CAS: 5505-63-5 Molecular Formula: C6 H13 N O5 . Cl H Molecular Weight (g/mol): 215.63 IUPAC Name: (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride SMILES: Cl.N[C@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO