missing translation for 'onlineSavingsMsg'
Learn More

β-D-Glucose, MP Biomedicals

Product Code. 11324189 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 11324189

Brand: MP Biomedicals 0210095391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • β-D-Glucose is a main source of energy for living organisms
  • It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-61-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063989
InChI Key WQZGKKKJIJFFOK-VFUOTHLCSA-N
Synonym beta-d-glucose, beta-d-glucopyranose, glucoside, b-d-glucopyranose, beta-glucose, beta-dextrose, .beta.-d-glucopyranose, b-glucose, b-d-glucose, unii-j4r00m814d
PubChem CID 64689
ChEBI CHEBI:15903
IUPAC Name (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 148°C to 155°C (literature)
Color White
Boiling Point ∼410.8°C (760 mmHg (Predicted) (literature))
Linear Formula C6H12O6
Quantity 1 kg
Solubility Information Soluble in water (1/20-clear solution) and alcohol (slightly).
Formula Weight 180.16
Percent Purity ≥80%
Physical Form Crystalline Powder
Chemical Name or Material β-D-GLUCOSE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.