accessibility menu, dialog, popup

UserName

CAS RN 492-61-5

HO(R)O(R)OH(R)OH(S)HO(S)HO

CAS RN 492-61-5

IUPAC Name: (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: β-D-glucose glucoside
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: WQZGKKKJIJFFOK-VFUOTHLCSA-N
SMILES: OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)