Learn More
R&D Systems™ Cyno Monkey Integrin beta 5 (XP_005547990) VersaClone cDNA
Specifications
Specifications
| Accession Number | XP_005547990 |
| Gene ID (Entrez) | 3693 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |