missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyltrichlorosilane, 97%

Product Code. p-7046817
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399165 25 g 25g
11309175 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399165 Supplier Thermo Scientific Alfa Aesar Supplier No. L16441.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14579-03-4 | C5H9Cl3Si | 203.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14579-03-4
Molecular Formula C5H9Cl3Si
Molecular Weight (g/mol) 203.562
MDL Number MFCD00236508
InChI Key FCMZRNUHEXJWGB-UHFFFAOYSA-N
Synonym cyclopentyltrichlorosilane, trichloro cyclopentyl silane, silane, trichlorocyclopentyl, cyclopentane, trichlorosilyl, trichlorosilylcyclopentane, cyclopentyl trichlorosilane, acmc-1c6jw, trichlorocyclopentylsilane
PubChem CID 84533
IUPAC Name trichloro(cyclopentyl)silane
SMILES C1CCC(C1)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 178°C to 179°C
Quantity 25 g
UN Number UN2987
Beilstein 2935230
Flash Point 68°C (154°F)
Formula Weight 203.57
Percent Purity 97%
Odor Acrid
Refractive Index 1.4705
Sensitivity Moisture sensitive
Density 1.226
Chemical Name or Material Cyclopentyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.