CAS RN 14579-03-4
IUPAC Name:
trichloro(cyclopentyl)silane
Synonyms:
trichloro(cyclopentyl)silane
Molecular Weight (g/mol):
203.56
Molecular Formula:
C5H9Cl3Si
InChi Key:
FCMZRNUHEXJWGB-UHFFFAOYSA-N
SMILES:
Cl[Si](Cl)(Cl)C1CCCC1
1
–
1
of
1
results
1
Cyclopentyltrichlorosilane, 97%
CAS: 14579-03-4 Molecular Formula: C5H9Cl3Si Molecular Weight (g/mol): 203.562 MDL Number: MFCD00236508 InChI Key: FCMZRNUHEXJWGB-UHFFFAOYSA-N Synonym: cyclopentyltrichlorosilane,trichloro cyclopentyl silane,silane, trichlorocyclopentyl,cyclopentane, trichlorosilyl,trichlorosilylcyclopentane,cyclopentyl trichlorosilane,acmc-1c6jw,trichlorocyclopentylsilane PubChem CID: 84533 IUPAC Name: trichloro(cyclopentyl)silane SMILES: C1CCC(C1)[Si](Cl)(Cl)Cl