missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl-1-cyclohexenyl Ketone, TRC

Product Code. 30356367
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356367 250 mg -
30354916 2.5 g 2.5g
30351026 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356367 Supplier Toronto Research Chemicals Supplier No. TRCC988027250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74598-74-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.3
Synonym 1-Cyclohexen-1-yl cyclohexyl, 1-Cyclohexen-1-ylcyclohexylmethanone, Cyclohex-1-enyl cyclohexyl ketone
IUPAC Name 1-(cyclohexen-1-yl)cyclohexane-1-carbaldehyde
SMILES O=C(C1CCCCC1)C2=CCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H20O/c14-11-13(9-5-2-6-10-13)12-7-3-1-4-8-12/h7,11H,1-6,8-10H2
Formula Weight 192.1514
Chemical Name or Material Cyclohexyl-1-cyclohexenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.