CAS RN 74598-74-6
CAS RN 74598-74-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclohexyl-1-cyclohexenyl Ketone, TRC
CAS: 74598-74-6 Molecular Formula: C13H20O Molecular Weight (g/mol): 192.3 Synonym: 1-Cyclohexen-1-yl cyclohexyl,1-Cyclohexen-1-ylcyclohexylmethanone,Cyclohex-1-enyl cyclohexyl ketone IUPAC Name: 1-(cyclohexen-1-yl)cyclohexane-1-carbaldehyde SMILES: O=C(C1CCCCC1)C2=CCCCC2