Learn More
MedChemExpress Cucurbitacin I
Product Code. 16481628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481628 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16481628 Supplier MedChemExpress Supplier No. HYN1405
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Cucurbitacin I is a natural selective inhibitor of JAK2/STAT3, with potent anti-cancer activity.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 514.65 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 2222-07-3
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 100 mg/mL (194.31 mM)
Synonym Elatericin B∣ JSI-124∣ NSC-521777
Target STAT∣JAK
Quantity 5 mg