Learn More
Corning™ Interleukin-2 (IL-2), Human Recombinant
Product Code. 10284421 Shop All Corning Products
Change view
Click to view available options
Quantity:
10,000 U
50,000 U
Unit Size:
10000 units
50000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10284421 10,000 U 10000 units
10010831 50,000 U 50000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10284421 Supplier Corning™ Supplier No. 354043
  • ONLINE EXCLUSIVE PRICE
    245.62 EUR
    233.41 EUR / Pack of 10000
    Save 12.21 EUR
    4% Off
 Restricted Item
Estimated Shipment: 24-02-2027
Elicit a range of responses from many cell types
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Cytokines are a class of signaling molecules or intercellular mediators (proteins, peptides, and glycoproteins) that primarily affect cells of the immune system; however they can affect various cell types outside of the immune system. They differ from classical hormones in that they are produced by a number of tissue or cell types rather than by specialized glands. The effects of cytokines on cells can yield different outcomes. Some cytokines cause cell proliferation, while others may cause chemotaxis between cell types, and others can even cause cell death. Cytokines and growth factors are similar in their structure and mechanism of action. Both bind and initiate signaling pathways and many share several intracellular signaling components.
Corning offers a range of cytokines and glycoproteins that are derived from different species. These cytokines have been used to promote long-term proliferation in cell culture and to induce cell-mediated immune responses in a variety of cell types.
1Biological Response Modifiers Program Jurkat IL-2 reference reagent.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Frozen in 55 mm Sodium Phosphate Buffer, containing 53 mm Sucrose and 0.4% HAS
Product Type IL-2
Purity >96% by SDS-PAGE analysis
Quantity 10,000 U
Source Recombinant E. coli
Sterility Sterile
Content And Storage -70°C
Reactivity Human