Learn More
Colupulone, TRC
Product Code. 30364284
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364284 50 mg 50mg
30349018 25 mg -
30360006 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30364284 Supplier Toronto Research Chemicals Supplier No. TRCC21475550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 468-27-9
Molecular Formula C25 H36 O4
Molecular Weight (g/mol) 400.55
Synonym Colupulone A, Colupulone I
IUPAC Name 3,5-dihydroxy-4,6,6-tris(3-methylbut-2-enyl)-2-(2-methylpropanoyl)cyclohexa-2,4-dien-1-one
SMILES CC(C)C(=O)C1=C(O)C(=C(O)C(CC=C(C)C)(CC=C(C)C)C1=O)CC=C(C)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C25H36O4/c1-15(2)9-10-19-22(27)20(21(26)18(7)8)24(29)25(23(19)28,13-11-16(3)4)14-12-17(5)6/h9,11-12,18,27-28H,10,13-14H2,1-8H3
Formula Weight 400.2614
Chemical Name or Material Colupulone