CAS RN 468-27-9
CAS RN 468-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Colupulone, TRC
CAS: 468-27-9 Molecular Formula: C25 H36 O4 Molecular Weight (g/mol): 400.55 Synonym: Colupulone A,Colupulone I IUPAC Name: 3,5-dihydroxy-4,6,6-tris(3-methylbut-2-enyl)-2-(2-methylpropanoyl)cyclohexa-2,4-dien-1-one SMILES: CC(C)C(=O)C1=C(O)C(=C(O)C(CC=C(C)C)(CC=C(C)C)C1=O)CC=C(C)C