Learn More
Clomethiazolum, TRC
Product Code. 30010376
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30010376 Supplier Toronto Research Chemicals Supplier No. TRCC587020100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 533-45-9
Molecular Formula C6 H8 Cl N S
Molecular Weight (g/mol) 161.65
Synonym Thiazole, 5-(2-chloroethyl)-4-methyl-, 5-(2-Chloroethyl)-4-methylthiazole, 4-Methyl-5-(β-chloroethyl)thiazole, 5-(β-Chloroethyl)-4-methylthiazole, Chlorethiazol, Chlorethiazole, Chlormethiazol, Chlormethiazole, Clomethiazole, Clomethiazolum, Distraneurin, Emineurina, Somnevrin
IUPAC Name 5-(2-chloroethyl)-4-methyl-1,3-thiazole
SMILES Cc1ncsc1CCCl
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H8ClNS/c1-5-6(2-3-7)9-4-8-5/h4H,2-3H2,1H3
Formula Weight 161.0066
Chemical Name or Material Clomethiazole