CAS RN 533-45-9
CAS RN 533-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clomethiazolum, TRC
CAS: 533-45-9 Molecular Formula: C6 H8 Cl N S Molecular Weight (g/mol): 161.65 Synonym: Thiazole, 5-(2-chloroethyl)-4-methyl-,5-(2-Chloroethyl)-4-methylthiazole,4-Methyl-5-(β-chloroethyl)thiazole,5-(β-Chloroethyl)-4-methylthiazole,Chlorethiazol,Chlorethiazole,Chlormethiazol,Chlormethiazole,Clomethiazole,Clomethiazolum,Distraneurin,Emineurina,Somnevrin IUPAC Name: 5-(2-chloroethyl)-4-methyl-1,3-thiazole SMILES: Cc1ncsc1CCCl