missing translation for 'onlineSavingsMsg'
Learn More

cis-Zeatin, MP Biomedicals™

Product Code. 11498511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498511 1 mg 1mg
11408521 5 mg 5mg
11418521 10 mg 10mg
11428521 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498511 Supplier MP Biomedicals Supplier No. 0215227101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 32771-64-5
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.248
MDL Number MFCD00070217
InChI Key UZKQTCBAMSWPJD-UQCOIBPSSA-N
Synonym cis-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, z-2-methyl-4-1h-purin-6-ylamino but-2-en-1-ol, z-4-7h-purin-6-yl amino-2-methylbut-2-en-1-ol, 6-4-hydroxy-3-methyl-cis-2-butenylamino purine, e-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino-2-buten-1-ol, cis-zeatin hplc, z-2-methyl-4-7h-purin-6-ylamino but-2-en-1-ol
PubChem CID 688597
ChEBI CHEBI:46570
IUPAC Name (Z)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Quantity 1 mg
Formula Weight 219.2
Physical Form Solid
Chemical Name or Material cis-Zeatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.