CAS RN 32771-64-5
IUPAC Name:
2-methyl-4-[(7H-purin-6-yl)amino]but-2-en-1-ol
Synonyms:
zeatin
Molecular Weight (g/mol):
219.25
Molecular Formula:
C10H13N5O
InChi Key:
UZKQTCBAMSWPJD-UHFFFAOYSA-N
SMILES:
CC(CO)=CCNC1=C2NC=NC2=NC=N1
1
–
1
of
1
results
1
cis-Zeatin, MP Biomedicals™
CAS: 32771-64-5 Molecular Formula: C10H13N5O Molecular Weight (g/mol): 219.248 MDL Number: MFCD00070217 InChI Key: UZKQTCBAMSWPJD-UQCOIBPSSA-N Synonym: cis-zeatin,2z-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol,z-2-methyl-4-1h-purin-6-ylamino but-2-en-1-ol,z-4-7h-purin-6-yl amino-2-methylbut-2-en-1-ol,6-4-hydroxy-3-methyl-cis-2-butenylamino purine,e-zeatin,2z-2-methyl-4-9h-purin-6-ylamino-2-buten-1-ol,cis-zeatin hplc,z-2-methyl-4-7h-purin-6-ylamino but-2-en-1-ol PubChem CID: 688597 ChEBI: CHEBI:46570 IUPAC Name: (Z)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol SMILES: CC(=CCNC1=NC=NC2=C1NC=N2)CO