missing translation for 'onlineSavingsMsg'
Learn More

Cinnamic Acid, TRC

Product Code. 30355427
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355427 10 g 10g
30351928 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355427 Supplier Toronto Research Chemicals Supplier No. TRCC44203010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-82-9
Molecular Formula C9 H8 O2
Molecular Weight (g/mol) 148.159
Synonym 3-Phenyl-2-propenoic Acid, 3-Phenylacrylic Acid, NSC 623441, NSC 9189, Phenylacrylic Acid, β-Phenylacrylic Acid
IUPAC Name 3-phenylprop-2-enoic acid
SMILES OC(=O)C=Cc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)
Formula Weight 148.052
Chemical Name or Material Cinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.