CAS RN 621-82-9
CAS RN 621-82-9
IUPAC Name:
(2E)-3-phenylprop-2-enoic acid
Synonyms:
cinnamic acid (z)-cinnamic acid cinnamic acid (natural) cinnamic acid, (Z)- cinnamic acid, (z)- kyselina skoricove trans-zimtsaeure sodium salt cinnamic acid acidum cinnamylicum trans-3-phenylpropensaeure
Molecular Weight (g/mol):
148.16
Molecular Formula:
C9H8O2
InChi Key:
WBYWAXJHAXSJNI-VOTSOKGWSA-N
SMILES:
OC(=O)\C=C\C1=CC=CC=C1
1
–
2
of
2
results
1
Cinnamic Acid, TRC
CAS: 621-82-9 Molecular Formula: C9 H8 O2 Molecular Weight (g/mol): 148.159 Synonym: 3-Phenyl-2-propenoic Acid,3-Phenylacrylic Acid,NSC 623441,NSC 9189,Phenylacrylic Acid,β-Phenylacrylic Acid IUPAC Name: 3-phenylprop-2-enoic acid SMILES: OC(=O)C=Cc1ccccc1