missing translation for 'onlineSavingsMsg'
Learn More

Chrysene, TRC

Product Code. 30133397
Change view
Click to view available options
Quantity:
25 mg
250 mg
5 g
Unit Size:
250mg
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133397 250 mg 250mg
30135596 25 mg 25mg
30008675 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133397 Supplier Toronto Research Chemicals Supplier No. TRCC428000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218-01-9
Molecular Formula C18 H12
Molecular Weight (g/mol) 228.29
Synonym 1,2-Benzophenanthrene, 1,2-Benzphenanthrene, Benzo[a]phenanthrene, NSC 6175, [4]Phenacene
IUPAC Name chrysene
SMILES c1ccc2c(c1)ccc3c4ccccc4ccc23

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H
Formula Weight 228.0939
Percent Purity >95%
Chemical Name or Material Chrysene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.