accessibility menu, dialog, popup

UserName

CAS RN 218-01-9

IUPAC Name: chrysene
Synonyms: chrysene benz(a)phenanthrene 1,2-benzphenanthrene ccris 161 rcra waste number U050 1,2,5,6-dibenzonaphthalene benzo(a)phenanthrene 1,2-benzophenanthrene
Molecular Weight (g/mol): 228.29
Molecular Formula: C18H12
InChi Key: WDECIBYCCFPHNR-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=C2C=CC2=CC=CC=C32)=C1
Molecular Weight (g/mol) 
  • (3)
  • (9)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (3)
  • (2)
  • (3)
  • (9)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)