Learn More
Proteintech ChromoTek GFP-Trap™ Agarose Kit
Product Code. 17323353
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. Quantity unitSize
17323353 20 Reactions 20 reactions
1 options
Product Code. 17323353
Brand: Proteintech gtak20
Kit including 20 reactions of GFP-Trap™ Agarose and lysis, dilution, wash, and elution buffers.
  • GFP-Trap™ Agarose is an affinity resin for IP of GFP-fusion proteins
  • Consists of a GFP Nanobody/ VHH coupled to agarose beads
  • The ChromoTek GFP-Trap™ Agarose is a ready-to-use reagent
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions