Learn More
Chloroiodoacetic Acid, TRC
Product Code. 30134451
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134451 100 mg 100mg
18071210 10 mg 10mg
18012710 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134451 Supplier Toronto Research Chemicals Supplier No. TRCC367170100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53715-09-6
Molecular Formula C2 H2 Cl I O2
Molecular Weight (g/mol) 220.39
Synonym 2-Chloro-2-iodoacetic acid (ACI), Acetic acid, chloroiodo- (6CI, 9CI), Chloroiodoacetic acid
IUPAC Name 2-chloro-2-iodoacetic acid
SMILES OC(=O)C(Cl)I
Specifikationer
Quantity 100 mg
InChI Formula InChI=1S/C2H2ClIO2/c3-1(4)2(5)6/h1H,(H,5,6)
Formula Weight 219.8788
Chemical Name or Material Chloroiodoacetic Acid
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.