CAS RN 53715-09-6
CAS RN 53715-09-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chloroiodoacetic Acid, TRC
CAS: 53715-09-6 Molecular Formula: C2 H2 Cl I O2 Molecular Weight (g/mol): 220.39 Synonym: 2-Chloro-2-iodoacetic acid (ACI),Acetic acid, chloroiodo- (6CI, 9CI),Chloroiodoacetic acid IUPAC Name: 2-chloro-2-iodoacetic acid SMILES: OC(=O)C(Cl)I