Learn More
Chloroacetic Acid, Sigma-Aldrich™
Product Code. 15878260
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
50 kg
Unit Size:
100g
1kg
2.5kg
50kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878260 50 kg 50kg
15858260 100 g 100g
15876230 1 kg 1kg
15868260 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15878260 Supplier Sigma Aldrich Supplier No. 8004129050
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-11-8
Specifications
Melting Point 63°C
Density 1.64 g/cm3 at 20°C
MDL Number MFCD00002683
Quantity 50 kg
Synonym Chloroacetic acid, Monochloroacetic acid, Chloroacetic acid
Molecular Weight (g/mol) 94.5
Grade Research