accessibility menu, dialog, popup

UserName

OHOCl

CAS RN 79-11-8

IUPAC Name: 2-chloroacetic acid
Synonyms: chloroacetic acid chloroacetic acid, calcium (3:1) salt temmler brand of chloroacetic acid potassium salt chloroacetic acid epa pesticide chemical code 279400 chloroacetic acid preparation acide chloroacetique chloroacetic acid, sodium (2:1) salt acide monochloracetique silver salt chloroacetic acid
Molecular Weight (g/mol): 94.49
Molecular Formula: C2H3ClO2
InChi Key: FOCAUTSVDIKZOP-UHFFFAOYSA-N
SMILES: OC(=O)CCl
Quantity 
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (15)
Boiling Point 
  • (5)
  • (9)
  • (3)
Percent Purity 
  • (2)
  • (9)
  • (5)
Physical Form 
  • (3)
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (1)
  • (9)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (15)
  • (5)
  • (9)
  • (3)
  • (2)
  • (9)
  • (5)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)