missing translation for 'onlineSavingsMsg'
Learn More

CEACAM5/CD66e Antibody (308/3-3), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18214884 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214884 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18214884 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP254624JF549
  • ONLINE EXCLUSIVE PRICE
    456.00 EUR
    430.50 EUR / 0.1mL
    Save 25.50 EUR
    5% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunoprecipitation, Sandwich ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM5/CD66e
Applications CyTOF, ELISA, Flow Cytometry, Immunoprecipitation, Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 308/3-3
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Gene Symbols CEACAM5
Host Species Mouse
Immunogen Recombinant soluble human CEACAM3-Fc produced in HEK293 cells
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1048
Test Specificity Clone 308/3-3 reacts with human CEACAM3 and CEACAM5, but not human CEACAM1/4/6/7/8, rat CEACAM1, or mouse CEACAM1 & CEACAM2.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.