missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD90.2 Monoclonal specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | CD90.2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 53-2.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Formulation | Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide |
| Gene Alias | CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen |
| Gene Symbols | THY1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?