missing translation for 'onlineSavingsMsg'
Learn More

CD34 Rabbit anti-Human, Rat, Clone-HPCA1/1806R, Novus Biologicals™

Product Code. 18214630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18214630 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214630 Supplier Novus Biologicals Supplier No. NBP254355100UG
  • ONLINE EXCLUSIVE PRICE
    661.00 EUR
    624.75 EUR / 100µg
    Save 36.25 EUR
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD34 Monoclonal specifically detects CD34 in Human, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34
Applications CyTOF, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HPCA1/1806R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Gene Symbols CD34
Host Species Rabbit
Immunogen Recombinant full-length human HPCA1 protein
Molecular Weight of Antigen 95 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human, Rat
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.