missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Antibody (OKT3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
205.80 EUR - 373.00 EUR
Specifications
| Antigen | CD3 |
|---|---|
| Clone | OKT3 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot reported in scientific literature (PMID 28699640), Flow Cytometry, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay, In vivo assay |
| Applications | Western Blot, Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18053834
|
Novus Biologicals
NBP2-25186SS |
0.025 mg |
218.00 EUR 205.80 EUR / 25µg Save 12.20 EUR 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795313
|
Novus Biologicals
NBP2-25186 |
373.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Functional, Immunoblotting, In vitro assay, In vivo assay.Specifications
| CD3 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
| NP_000723 | |
| 916 | |
| Human PBMCs were used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 23.1 kDa |
| OKT3 | |
| Western Blot reported in scientific literature (PMID 28699640), Flow Cytometry, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay, In vivo assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| CD3 | |
| IgG2a κ | |
| Protein G purified | |
| This product reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title