Learn More
CD3 Antibody (OKT3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand:  Novus Biologicals NBP2-25186
 View more versions of this product
Product Code. 18795313
  • 373.00 EUR / 0.10mg
Description
CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Functional, Immunoblotting, In vitro assay, In vivo assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Functional Assay, Immunoblot
Mouse
Purified
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ
Certificates
Promotions
Promotions
For Research Use Only