missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Functional, Immunoblotting, In vitro assay, In vivo assay.
Specifications
Specifications
| Antigen | CD3 |
| Applications | Western Blot, Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay |
| Classification | Monoclonal |
| Clone | OKT3 |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot reported in scientific literature (PMID 28699640), Flow Cytometry, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay, In vivo assay |
| Gene Accession No. | NP_000723 |
| Gene Alias | CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE |
| Gene Symbols | CD3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?