Learn More
Bochem™ Thermo Scientific™ Cast Iron Chromium Plated Standard Retort Clamp
Product Code. 10363571
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10363571 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10363571 Supplier Bochem™ Supplier No. 5462
  • ONLINE EXCLUSIVE PRICE
    17.30 EUR
    13.12 EUR / Pack of 1
    Save 4.18 EUR
    24% Off
Estimated Shipment: 26-11-2026
Hold test tubes and other glassware up to 40mm wide. BOCHEM™ Thermo Scientific™ Cast Iron Chromium Plated Standard Retort Clamp serves a variety of functions in the modern laboratory.
  • Malleable clast iron
  • Chromium plated
  • Green
  • With cork inlay
  • Round/round
  • DIN 12894
Specifications
Diameter (Metric) Inner 40 mm
Material Cast Iron Chromium Plated
Quantity 1 Each
Type Tools and Accessories