missing translation for 'onlineSavingsMsg'
Learn More

Caspase-8 Antibody (3B10), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18215066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215066 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215066 Supplier Novus Biologicals Supplier No. NBP280098AF750
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody

Caspase-8 Monoclonal specifically detects Caspase-8 in Mouse samples. It is validated for ELISA/Flow Cytometry/Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-8
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3B10
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Rat
Immunogen Recombinant mouse caspase-8 (p20 subunit).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Apoptosis Detections, Cancer, Caspases, Cell Biology, Necroptosis, Phospho Specific, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 841
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.