missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Carvedilol
GREENER_CHOICE

Product Code. 30001418
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001418 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001418 Supplier Tocris Bioscience™ Supplier No. 2685/50
  • ONLINE EXCLUSIVE PRICE
    169.00 EUR
    159.60 EUR / 50mg
    Save 9.40 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and non-selective β antagonist; also potent α1 antagonist

Potent β-adrenoceptor and α1-adrenoceptor antagonist (Ki values are 0.81, 0.96 and 2.2 nM for β1-, β2- and α1-adrenoceptors respectively) that displays antihypertensive and peripheral vasodilatory activity. Blocks cardiac inward-rectifier K+ (KIR) channels, voltage-dependent Ca2+ channels and exhibits antioxidant properties at higher concentrations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 72956-09-3
Quantity 50 mg
Molecular Formula C24H26N2O4
Formula Weight 406.47
Purity 98%
Molecular Weight (g/mol) 406.48
Chemical Name or Material Carvedilol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.