Learn More
Cardanol (Mixture) (>80%), TRC
Product Code. 30010048
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010048 10 g 10g
30003972 5 g 5g
30006120 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010048 Supplier Toronto Research Chemicals Supplier No. TRCC18338310G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37330-39-5
Molecular Formula C84 H132 O4
Molecular Weight (g/mol) 1205.98
Synonym Card-phenol, Cardolite NC 700, EJ-C 513, GX 2512, Lite 2013, NX 2021
IUPAC Name 3-((8Z,11Z)-pentadeca-8,11,14-trien-1-yl)phenol and 3-((8Z,11Z)-pentadeca-8,11-dien-1-yl)phenol and (Z)-3-(pentadec-8-en-1-yl)phenol and 3-pentadecylphenol
SMILES C=CC/C=C\C/C=C\CCCCCCCC1=CC(O)=CC=C1.CCC/C=C\C/C=C\CCCCCCCC2=CC(O)=CC=C2.OC3=CC=CC(CCCCCCCCCCCCCCC)=C3.OC4=CC=CC(CCCCCCC/C=C\CCCCCC)=C4
Specifications
Quantity 10 g
InChI Formula InChI=1S/C21H36O.C21H34O.C21H32O.C21H30O/c4*1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-20-17-15-18-21(22)19-20/h15,17-19,22H,2-14,16H2,1H3;7-8,15,17-19,22H,2-6,9-14,16H2,1H3;4-5,7-8,15,17-19,22H,2-3,6,9-14,16H2,1H3;2,4-5,7-8,15,17-19,22H,1,3,6,9-14,16H2/b;8-7-;2*5-4-,8-7-
Formula Weight 1205.01
Chemical Name or Material Cardanol (Mixture)