CAS RN 37330-39-5
CAS RN 37330-39-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cardanol (Mixture) (>80%), TRC
CAS: 37330-39-5 Molecular Formula: C84 H132 O4 Molecular Weight (g/mol): 1205.98 Synonym: Card-phenol,Cardolite NC 700,EJ-C 513,GX 2512,Lite 2013,NX 2021 IUPAC Name: 3-((8Z,11Z)-pentadeca-8,11,14-trien-1-yl)phenol and 3-((8Z,11Z)-pentadeca-8,11-dien-1-yl)phenol and (Z)-3-(pentadec-8-en-1-yl)phenol and 3-pentadecylphenol SMILES: C=CC/C=C\C/C=C\CCCCCCCC1=CC(O)=CC=C1.CCC/C=C\C/C=C\CCCCCCCC2=CC(O)=CC=C2.OC3=CC=CC(CCCCCCCCCCCCCCC)=C3.OC4=CC=CC(CCCCCCC/C=C\CCCCCC)=C4