missing translation for 'onlineSavingsMsg'
Learn More

Carbofuranphenol-3-keto, TRC

Product Code. 30010552
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010552 5 mg 5mg
30007498 1 mg 1mg
30003914 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010552 Supplier Toronto Research Chemicals Supplier No. TRCC1765985MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17781-16-7
Molecular Formula C10 H10 O3
Molecular Weight (g/mol) 178.18
Synonym 3(2H)-Benzofuranone, 7-hydroxy-2,2-dimethyl-, 7-Hydroxy-2,2-dimethyl-3(2H)-benzofuranone, 2,2-Dimethyl-3-oxo-2,3-dihydro-1-benzofuran-7-ol, 2,3-Dihydro-2,2-dimethyl-3-keto-7-hydroxybenzofuran, 2,3-Dihydro-2,2-dimethyl-3-oxo-7-benzofuranol, 2,3-Dihydro-7-hydroxy-2,2-dimethyl-3-ketobenzofuran, 2,3-Dihydro-7-hydroxy-2,2-dimethyl-3-oxobenzofuran, 3-Ketocarbofuran phenol, 7-Hydroxy-2,2-dimethylbenzofuran-3-one, Carbofuranphenol-3-keto
IUPAC Name 7-hydroxy-2,2-dimethyl-1-benzofuran-3-one
SMILES CC1(C)Oc2c(O)cccc2C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H10O3/c1-10(2)9(12)6-4-3-5-7(11)8(6)13-10/h3-5,11H,1-2H3
Formula Weight 178.063
Chemical Name or Material Carbofuranphenol-3-keto
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.