CAS RN 17781-16-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carbofuranphenol-3-keto, TRC
CAS: 17781-16-7 Molecular Formula: C10 H10 O3 Molecular Weight (g/mol): 178.18 Synonym: 3(2H)-Benzofuranone, 7-hydroxy-2,2-dimethyl-,7-Hydroxy-2,2-dimethyl-3(2H)-benzofuranone,2,2-Dimethyl-3-oxo-2,3-dihydro-1-benzofuran-7-ol,2,3-Dihydro-2,2-dimethyl-3-keto-7-hydroxybenzofuran,2,3-Dihydro-2,2-dimethyl-3-oxo-7-benzofuranol,2,3-Dihydro-7-hydroxy-2,2-dimethyl-3-ketobenzofuran,2,3-Dihydro-7-hydroxy-2,2-dimethyl-3-oxobenzofuran,3-Ketocarbofuran phenol,7-Hydroxy-2,2-dimethylbenzofuran-3-one,Carbofuranphenol-3-keto IUPAC Name: 7-hydroxy-2,2-dimethyl-1-benzofuran-3-one SMILES: CC1(C)Oc2c(O)cccc2C1=O